Learn More
Aviva Systems Biology Corp PDCD2L Antibody - N-terminal region (ARP50714_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50714_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDCD2L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PDCD2L. Alternate names: MGC13096ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60417