Learn More
Aviva Systems Biology Corp PDCD4 Antibody - N-terminal region (AVARP09045_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP09045_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PDCD4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PDCD4. Alternate names: H731ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62585