Learn More
Aviva Systems Biology Corp PDCD7 Antibody - N-terminal region (ARP58776_P050) | 100uL
SDP
Catalog No. 5001462421
Encompass_Preferred
This is a rabbit polyclonal antibody against PDCD7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PDCD7. Alternate names: ES18, HES18, MGC22015ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig
Catalog No. 50-014-62421 Supplier Aviva Systems Biology Corp Supplier No. ARP58776_P050
This is a rabbit polyclonal antibody against PDCD7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PDCD7. Alternate names: ES18, HES18, MGC22015ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig