Learn More
Aviva Systems Biology Corp PDCD8 Antibody - N-terminal region (AVARP00028_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP00028_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDCD8. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human PDCD8. Alternate names: RP3-438D16.2, AIF, MGC111425, PDCD8, COXPD6ApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Sheep
Catalog No. 50-014-62456