Learn More
CPC Scientific H-Gln-Thr-Glu-Lys-Met-Trp-Gln-Arg-Leu-Lys-Gly-Ile-Leu-Arg-Ser-Leu-Val-Lys-Gln-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific SIGN012A
Encompass
This peptide is a calmodulin binding domain. The isolated peptide in solution is unstructured, but it adopts the conformation of an amphipathic α-helix when it binds to CaM-Ca2+. ONE-LETTER SEQUENCE: QTEKMWQRLKGILRSLVKQMOLECULAR FORMULA: C104H180N32O27S1MOLECULAR WEIGHT:2740.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: REFERENCES: Frederick, K. et al. Nature 448, 325 (2007); André. I. et al. Biophys. J. 90, 2903 (2006)
Catalog No. 50-211-0407