Learn More
Medchemexpress LLC HY-101490 5mg Medchemexpress, PDE1-IN-2 CAS:1904611-63-7 Purity:>98%
SDP
Catalog No. 501932149
Encompass_Preferred
Medchemexpress, HY-101490 5mg PDE1-IN-2 CAS:1904611-63-7 PDE1-IN-2 is an inhibitor of PDE1 extracted from patent WO2016/55618 A1, example 31; has IC 50 values of 6, 140 and 164 nM for PDE1C, PDE1B and PDE1A, respectvely. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2149 Supplier Medchemexpress LLC Supplier No. HY1014905MG
Medchemexpress, HY-101490 5mg PDE1-IN-2 CAS:1904611-63-7 PDE1-IN-2 is an inhibitor of PDE1 extracted from patent WO2016/55618 A1, example 31; has IC 50 values of 6, 140 and 164 nM for PDE1C, PDE1B and PDE1A, respectvely. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.