Learn More
Medchemexpress LLC PDE12-IN-1 | 2259620-80-7 | 98.9% | 616.48 | C31H27BrFN5O3 | 10 MG
SDP
Catalog No. 5000226761
Encompass_Preferred
PDE12-IN-1 is a selective phosphodiesterase 12 (PDE12) inhibitor provided for laboratory research use. It shows potent enzyme inhibition and modulates 2′,5′-linked adenylate polymer levels in cellular assays. Intended for in vitro biochemical and cell-based studies; not for human or clinical use.
  • Selective PDE12 inhibition with reported pIC50 ≈ 9.1.
  • Suitable for biochemical and cellular assays.
  • High purity: 98.9%.
  • Available in small pack sizes for screening and optimization.
  • Molecular formula C31H27BrFN5O3; molecular weight 616.48.
Catalog No. 50-002-26761 Supplier Medchemexpress LLC Supplier No. HY11731810MG
PDE12-IN-1 is a selective phosphodiesterase 12 (PDE12) inhibitor provided for laboratory research use. It shows potent enzyme inhibition and modulates 2′,5′-linked adenylate polymer levels in cellular assays. Intended for in vitro biochemical and cell-based studies; not for human or clinical use.
  • Selective PDE12 inhibition with reported pIC50 ≈ 9.1.
  • Suitable for biochemical and cellular assays.
  • High purity: 98.9%.
  • Available in small pack sizes for screening and optimization.
  • Molecular formula C31H27BrFN5O3; molecular weight 616.48.