Learn More
Aviva Systems Biology Corp PDE1A Antibody - C-terminal region (ARP53625_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53625_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDE1A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human PDE1A. Alternate names: HCAM1, HSPDE1A, MGC26303ApplicationWB. Species reactivity: Dog, Human, Mouse, Rat
Catalog No. 50-285-859