Learn More
Sino Biological Recombinant Human PTP sigma/PTPRS Protein (GST Tag), Active
Catalog No. 502573577
Encompass_Preferred
Recombinant human PTPRS (883-1210) was expressed in E.coli cells using a N-terminal GST tag.
Catalog No. 50-257-3577 Supplier Sino Biological Supplier No. P6421G50
Recombinant human PTPRS (883-1210) was expressed in E.coli cells using a N-terminal GST tag.