Learn More
Medchemexpress LLC PDE2A-IN-3 | 1426833-08-0 | 295.73 | 50 MG
SDP
Catalog No. 5000447283
Encompass_Preferred
PDE2A-IN-3 (Example Ic1) is a selective phosphodiesterase 2A (PDE2A) inhibitor with IC50s of 69 nM and 1762 nM for PDE2A and PDE10, respectively. It can be used for the study of neurological and psychiatric disorders.
  • Selective phosphodiesterase 2A (PDE2A) inhibitor
  • IC50 of 69 nM for PDE2A and 1762 nM for PDE10
  • Useful for research into neurological and psychiatric disorders
Catalog No. 50-004-47283 Supplier Medchemexpress LLC Supplier No. HYU0042750MG
PDE2A-IN-3 (Example Ic1) is a selective phosphodiesterase 2A (PDE2A) inhibitor with IC50s of 69 nM and 1762 nM for PDE2A and PDE10, respectively. It can be used for the study of neurological and psychiatric disorders.
  • Selective phosphodiesterase 2A (PDE2A) inhibitor
  • IC50 of 69 nM for PDE2A and 1762 nM for PDE10
  • Useful for research into neurological and psychiatric disorders