Learn More
Aviva Systems Biology Corp PDE3A Antibody - N-terminal region (ARP44356_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44356_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDE3A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PDE3A. Alternate names: CGI-PDE, CGI-PDE A, CGI-PDE-AApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rat
Catalog No. 50-014-58787