Learn More
Medchemexpress LLC PDE3A protein, human (sf9, His-GST) | >90.0% | 10 UG
SDP
Catalog No. 5000292308
Encompass_Preferred
Recombinant human phosphodiesterase 3A (PDE3A) fragment expressed in Sf9 insect cells and supplied as an N-terminal His-GST fusion. Purity is >90% by reducing SDS-PAGE. Intended for use in biochemical assays, binding studies, and structural characterization; enzymatic activity testing is in progress.
  • Expressed in Sf9 insect cells.
  • N-terminal His-GST fusion for straightforward purification and detection.
  • Purity >90.0% as determined by reducing SDS-PAGE.
  • Available in small-scale packs such as 10 μg for assay development.
  • Suitable for biochemical enzymatic assays and protein interaction studies.
Catalog No. 50-002-92308 Supplier Medchemexpress LLC Supplier No. HYP7596110UG
Recombinant human phosphodiesterase 3A (PDE3A) fragment expressed in Sf9 insect cells and supplied as an N-terminal His-GST fusion. Purity is >90% by reducing SDS-PAGE. Intended for use in biochemical assays, binding studies, and structural characterization; enzymatic activity testing is in progress.
  • Expressed in Sf9 insect cells.
  • N-terminal His-GST fusion for straightforward purification and detection.
  • Purity >90.0% as determined by reducing SDS-PAGE.
  • Available in small-scale packs such as 10 μg for assay development.
  • Suitable for biochemical enzymatic assays and protein interaction studies.