Learn More
Aviva Systems Biology Corp PDE3B Antibody - middle region (ARP44359_P050) | 100uL
SDP
Catalog No. 5001458788
Encompass_Preferred
This is a rabbit polyclonal antibody against PDE3B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PDE3B. Alternate names: HcGIP1, cGIPDE1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58788 Supplier Aviva Systems Biology Corp Supplier No. ARP44359_P050
This is a rabbit polyclonal antibody against PDE3B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PDE3B. Alternate names: HcGIP1, cGIPDE1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish