Learn More
Enzo Life Sciences PDE4A (catalytic domain) (human), (recombinant) (20 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE5210020
Encompass_Preferred
Alternative name: Phosphodiesterase 4A. Formulation: Liquid. In 20mM Tris-HCl, pH 8.0, containing 150mM NaCl, 2mM DTT. Source: Produced in E. coli. Active catalytic domain of phosphodiesterase 4A. Long Term Storage: -80°C.
Catalog No. 50-201-2063