Learn More
Medchemexpress LLC PDE4B-IN-2 5mg | 915082-52-9 | 387.88 | C19H18ClN3O2S | 5 MG
SDP
Catalog No. 5000226166
Encompass_Preferred
An orally active, selective phosphodiesterase 4B (PDE4B) inhibitor with potent anti-inflammatory activity; shows an IC50 of 15 nM for PDE4B and 1.7 μM for PDE4D. Supplied as a white to off-white solid with high purity and provided with storage recommendations for long-term stability.
  • Selective PDE4B inhibition (IC50 15 nM).
  • Lower activity against PDE4D (IC50 1.7 μM), supporting selectivity.
  • High purity research grade material for in vitro and in vivo studies.
  • Stable solid form with defined storage conditions for extended shelf life.
  • Demonstrated anti-inflammatory effects in cellular and mouse models.
Catalog No. 50-002-26166 Supplier Medchemexpress LLC Supplier No. HY1156875MG
An orally active, selective phosphodiesterase 4B (PDE4B) inhibitor with potent anti-inflammatory activity; shows an IC50 of 15 nM for PDE4B and 1.7 μM for PDE4D. Supplied as a white to off-white solid with high purity and provided with storage recommendations for long-term stability.
  • Selective PDE4B inhibition (IC50 15 nM).
  • Lower activity against PDE4D (IC50 1.7 μM), supporting selectivity.
  • High purity research grade material for in vitro and in vivo studies.
  • Stable solid form with defined storage conditions for extended shelf life.
  • Demonstrated anti-inflammatory effects in cellular and mouse models.