Learn More
Aviva Systems Biology Corp PDE4C Antibody - C-terminal region (ARP64361_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64361_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDE4C. It was validated on Western Blot. . Alternate names: DPDE1, MGC126222ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-245