Learn More
Aviva Systems Biology Corp Pde4d Antibody - N-terminal region (ARP56672_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56672_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Pde4d. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 9630011N22Rik, Dpde3ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-189