Learn More
Aviva Systems Biology Corp PDE5A Antibody - C-terminal region (ARP63250_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63250_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDE5A. It was validated on Western Blot. . Alternate names: CGB-PDE, CN5A, PDE5, PDE5A1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-921