Learn More
Enzo Life Sciences PDE5A (human), (recombinant) (10µg)
SDP
Catalog No. 502013918
Encompass_Preferred
Alternative name: Phosphodiesterase 5A, cGMP-specific 3,5-cyclic phosphodiesterase, CGB-PDE. Purity: ≥95%. Formulation: Liquid. In 50mM triethanolamine, pH 7.4, containing 1mM DTT, 100mM NaCl, 20% glycerol. Source: Produced in Sf9 insect cells. Recombinant human PDE5A (phosphodiesterase 5A) is fused at the C-terminus to a His-tag. Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-3918 Supplier Enzo Life Sciences Supplier No. ENZPRT1260010
Alternative name: Phosphodiesterase 5A, cGMP-specific 3,5-cyclic phosphodiesterase, CGB-PDE. Purity: ≥95%. Formulation: Liquid. In 50mM triethanolamine, pH 7.4, containing 1mM DTT, 100mM NaCl, 20% glycerol. Source: Produced in Sf9 insect cells. Recombinant human PDE5A (phosphodiesterase 5A) is fused at the C-terminus to a His-tag. Produced in a baculovirus expression system. Long Term Storage: -80°C.