Learn More
Enzo Life Sciences PDE5A polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ALX210099C050
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 857-875 (CA857EQQEKMLINGESGQAKRN875) of human PDE5A (phosphodiesterase 5A). UniProt ID: O76074. Species reactivity: Mouse Bovine. Applications: WB. Recommended Dilutions: Western Blot (1:10,000) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Epitope-affinity purified. Formulation: Liquid. In PBS containing 30% glycerol and 0.02% sodium azide. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.
Catalog No. 50-200-8067