Learn More
Aviva Systems Biology Corp PDE6B Antibody - C-terminal region (ARP63580_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63580_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDE6B. It was validated on Western Blot. . Alternate names: CSNB3, CSNBAD2, PDEB, RP40, rd1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-020