Learn More
Aviva Systems Biology Corp PDE7A Antibody - C-terminal region (ARP64420_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64420_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDE7A. It was validated on Western Blot. . Alternate names: HCP1, PDE7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-288-260