Learn More
Aviva Systems Biology Corp PDE7B Antibody - middle region (ARP55364_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55364_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDE7B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PDE7B. Alternate names: MGC88256, bA472E5.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61479