Learn More
Aviva Systems Biology Corp PDE8A Antibody - C-terminal region (ARP57753_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57753_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDE8A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human PDE8A. Alternate names: FLJ16150, HsT19550ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-371