Learn More
Medchemexpress LLC PDE9-in-1 | 2305087-92-5 | 99.7% | 362.40 g·mol^-1 | C17H23FN6O2 | 25MG
SDP
Catalog No. 5000239457
Encompass_Preferred
PDE9-IN-1 is a potent, selective, and orally bioavailable inhibitor of phosphodiesterase-9A (PDE9A) with an IC50 of 8.7 nM. It is provided as a high-purity research chemical intended for biochemical and cellular assays, and for preclinical pharmacology studies investigating cGMP signaling and PDE9A-related pathways. Supporting documentation such as a datasheet, certificate of analysis, and safety data sheet are available from the manufacturer.
Catalog No. 50-002-39457 Supplier Medchemexpress LLC Supplier No. HY12613725MG
PDE9-IN-1 is a potent, selective, and orally bioavailable inhibitor of phosphodiesterase-9A (PDE9A) with an IC50 of 8.7 nM. It is provided as a high-purity research chemical intended for biochemical and cellular assays, and for preclinical pharmacology studies investigating cGMP signaling and PDE9A-related pathways. Supporting documentation such as a datasheet, certificate of analysis, and safety data sheet are available from the manufacturer.