Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pde9-in-1 | 2305087-92-5 | MFCD32062828 | 99.7% | 362.41 | C17H23FN6O2 | 10MG
SDP

Catalog No. 5000239460
Encompass_Preferred

PDE9-IN-1 is a potent, selective, and orally bioavailable phosphodiesterase-9A (PDE9A) inhibitor with an IC50 of 8.7 nM. It demonstrates strong selectivity across PDE family enzymes and has shown in vivo efficacy: oral dosing at 2.5 and 5.0 mg/kg daily for 21 days improved learning and memory in a unilateral common carotid artery occlusion mouse model. Supplied as a solid, it is intended for biochemical and preclinical pharmacology studies.

  • Molecular formula: C17H23FN6O2
  • Molecular weight: 362.41 g/mol
  • Purity: 99.7%
  • Solubility: soluble in DMSO (100 mg/mL)
  • Storage: powder -20°C (long term), in solvent -80°C (short term)
  • Target: phosphodiesterase-9A (PDE9A); IC50 8.7 nM
  • Form: solid
  • Applications: biochemical assays and preclinical in vivo testing

Catalog No. 50-002-39460 Supplier Medchemexpress LLC Supplier No. HY12613710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PDE9-IN-1 is a potent, selective, and orally bioavailable phosphodiesterase-9A (PDE9A) inhibitor with an IC50 of 8.7 nM. It demonstrates strong selectivity across PDE family enzymes and has shown in vivo efficacy: oral dosing at 2.5 and 5.0 mg/kg daily for 21 days improved learning and memory in a unilateral common carotid artery occlusion mouse model. Supplied as a solid, it is intended for biochemical and preclinical pharmacology studies.

  • Molecular formula: C17H23FN6O2
  • Molecular weight: 362.41 g/mol
  • Purity: 99.7%
  • Solubility: soluble in DMSO (100 mg/mL)
  • Storage: powder -20°C (long term), in solvent -80°C (short term)
  • Target: phosphodiesterase-9A (PDE9A); IC50 8.7 nM
  • Form: solid
  • Applications: biochemical assays and preclinical in vivo testing

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.