Learn More
TARGETMOL CHEMICALS INC PDE9-IN-S-C33 5MG
SDP
Catalog No. 502580523
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PDE9-IN-(S)-C33 ((S)-C33) is a potent and selective inhibitor of PDE9 (IC50 = 11 nM). PDE9-IN-(S)-C33 can be used for central nervous system diseases and diabetes research. purity: 98%
Catalog No. 50-258-0523 Supplier TARGETMOL CHEMICALS INC Supplier No. T28352
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PDE9-IN-(S)-C33 ((S)-C33) is a potent and selective inhibitor of PDE9 (IC50 = 11 nM). PDE9-IN-(S)-C33 can be used for central nervous system diseases and diabetes research. purity: 98%