Learn More
Medchemexpress LLC HY-126498 100mg Medchemexpress, PDEC-NB CAS:874302-76-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY126498100MG
Encompass_Preferred
Medchemexpress, HY-126498 100mg PDEC-NB CAS:874302-76-8 PDEC-NB is a disulfide cleavable linker used for the antibody-drug conjugate (ADC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1111