Learn More
Deltatrak Inc PDF Mult-Use Data Logger Cryo, Packaged individually
SDP
Catalog No. NC1730768
Encompass_Preferred
USB PDF Data Logger with special stainless steel probe allows temperature measurements from -200C to 40C for cryogenic applications such as liquid nitrogen tanks. This reusable logger has a built-in USB connector makes it easy to program and download the logger without special reading devices. No added software necessary. Probe Measurement Range (-200C to +40C) Operating Range (-20C to 50C) Temperature Resolution 0.1F/C Accuracy 0.5C from -196C to 40C Data Capacity 16,000 data points Battery Two AAA Lithium batteries Casing ABS plastic Certification CE Mark, NIST traceable, RoHS Compliant FAA Compliance RTCA/DO-160 Display LCD, Red LED Alarm Waterproof Rating IP64 with silicon cap on System Requirements Windows PC with current operating system, Adobe Reader Download Requirements Direct USB connection to PC
Catalog No. NC1730768 Supplier Deltatrak Inc Supplier No. 40565
USB PDF Data Logger with special stainless steel probe allows temperature measurements from -200C to 40C for cryogenic applications such as liquid nitrogen tanks. This reusable logger has a built-in USB connector makes it easy to program and download the logger without special reading devices. No added software necessary. Probe Measurement Range (-200C to +40C) Operating Range (-20C to 50C) Temperature Resolution 0.1F/C Accuracy 0.5C from -196C to 40C Data Capacity 16,000 data points Battery Two AAA Lithium batteries Casing ABS plastic Certification CE Mark, NIST traceable, RoHS Compliant FAA Compliance RTCA/DO-160 Display LCD, Red LED Alarm Waterproof Rating IP64 with silicon cap on System Requirements Windows PC with current operating system, Adobe Reader Download Requirements Direct USB connection to PC