Learn More
Deltatrak Inc PDF Reusable Logger BluntProbe, Packaged individually
SDP
Supplier:  Deltatrak Inc 40520
Encompass_Preferred
This reusable logger features a blunt tip probe for monitoring internal product temperature. Featuring a built-in USB connector, the unique feature of this unit is the on-board software, making it a plug-and-play logger allowing personnel in the field quick access to temperature reports. Probe Measurement Range -40F to 185F (-40C to 85C) Operating Range -40F to 122F (-40C to 50C) Temperature Resolution 0.1F/C Accuracy 1.0F from 14F to 77F (0.5C from -10C to 25C); 2.0F above 77F (1.0C above 25C) 2.0F below 14F (1.0C below -10C) Data Capacity 32,496 points Data Download Time 5 seconds typical for 3823 points Programmable Alarm Levels High & Low levels over entire range displayed on LCD Battery Two AAA Lithium batteries, LCD % remaining power indicator Certification CE Mark, NIST traceable, RoHS Compliant FAA Compliance RTCA/DO-160 Display LCD, Red LED Alarm IP64 with silicon cap on System Requirements Windows PC with current OS
Catalog No. NC1730743