Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PDGF-AA Human 100ug
SDP

Catalog No. 5000184953
Encompass_Preferred

Recombinant human PDGF-AA is a lyophilized platelet-derived growth factor for stimulating cell proliferation, migration, and survival in cell culture and biochemical assays. Supplied with lot-specific formulation details and a certificate of analysis, it is intended for research applications that require activation of PDGF signaling pathways.

  • Approximately 18 kDa molecular weight based on SDS-PAGE.
  • Expressed in E. coli and supplied as lyophilized material.
  • Endotoxin level below 1 EU/μg as determined by LAL assay.
  • Formulated in filtered buffers; refer to the lot certificate of analysis for exact composition.
  • Store at -20°C; after reconstitution, stable at 4°C for 1 week or frozen for long-term storage.
  • Reconstitute according to datasheet recommendations and avoid concentrations below 100 μg/mL in ddH2O.

Catalog No. 50-001-84953 Supplier Medchemexpress LLC Supplier No. HYP70598100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human PDGF-AA is a lyophilized platelet-derived growth factor for stimulating cell proliferation, migration, and survival in cell culture and biochemical assays. Supplied with lot-specific formulation details and a certificate of analysis, it is intended for research applications that require activation of PDGF signaling pathways.

  • Approximately 18 kDa molecular weight based on SDS-PAGE.
  • Expressed in E. coli and supplied as lyophilized material.
  • Endotoxin level below 1 EU/μg as determined by LAL assay.
  • Formulated in filtered buffers; refer to the lot certificate of analysis for exact composition.
  • Store at -20°C; after reconstitution, stable at 4°C for 1 week or frozen for long-term storage.
  • Reconstitute according to datasheet recommendations and avoid concentrations below 100 μg/mL in ddH2O.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.