Learn More
Sigma Aldrich Fine Chemicals Biosciences PDGf AA human Animal compo
Catalog No. 502906538
Encompass
Platelet derived growth factors (PDGfs) are disulfide-linked glycoprotein dimers consisting of two 12.0-13.5kDa polypeptide chains designated as PDGf-A and PDGf-B chains. There are three naturally occurring PDGfs PDGf-AA PDGf-BB and PDGf-AB. Recombinant human PDGf-AA is a 28.5kDa disulfide-linked homodimer of two A chains (250 total amino acids).
Catalog No. 50-290-6538 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SRP326810UG
Platelet derived growth factors (PDGfs) are disulfide-linked glycoprotein dimers consisting of two 12.0-13.5kDa polypeptide chains designated as PDGf-A and PDGf-B chains. There are three naturally occurring PDGfs PDGf-AA PDGf-BB and PDGf-AB. Recombinant human PDGf-AA is a 28.5kDa disulfide-linked homodimer of two A chains (250 total amino acids).