Learn More
Medchemexpress LLC PDGF-AA Rat P.past 50ug
SDP
Catalog No. 5000179489
Encompass_Preferred
PDGF-AA protein is a key growth factor that serves as an effective mitogen for mesenchymal cells coordinates embryonic development and affects cell proliferation migration survival and chemotaxis It plays an important role in alveolar septal formation gastrointestinal tract development interstitial cell maturation spermatogenesis oligodendrocyte development and myelination of the spinal cord and cerebellum PDGF-AA Protein Rat (P pastoris His) is the recombinant rat-derived PDGF-AA protein expressed by P pastoris with N-His labeled tag
Catalog No. 50-001-79489 Supplier Medchemexpress LLC Supplier No. HYP7372050UG
PDGF-AA protein is a key growth factor that serves as an effective mitogen for mesenchymal cells coordinates embryonic development and affects cell proliferation migration survival and chemotaxis It plays an important role in alveolar septal formation gastrointestinal tract development interstitial cell maturation spermatogenesis oligodendrocyte development and myelination of the spinal cord and cerebellum PDGF-AA Protein Rat (P pastoris His) is the recombinant rat-derived PDGF-AA protein expressed by P pastoris with N-His labeled tag