Learn More
Sigma Aldrich Fine Chemicals Biosciences Platelet-Derived Growth Factor-BB human PDGF-BB, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture | MFCD00213023 | 10UG
Catalog No. NC1912373
Encompass
Platelet-Derived Growth Factor-BB human PDGF-BB, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture | Purity: >=98% (HPLC and SDS-PAGE) | MFCD00213023 | 10UG
Catalog No. NC1912373 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P320110UG
Platelet-Derived Growth Factor-BB human PDGF-BB, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture | Purity: >=98% (HPLC and SDS-PAGE) | MFCD00213023 | 10UG