Learn More
Creative Biomart PDGF-BB
Catalog No. 5000332702
Encompass
PDGF-BB isomer is found to be critical in angiogenesis and is highly expressed in tumor mediating oncogenic pericyte recruitment PDGF-BB is an important regulator of bone homeostasis repair and regeneration Potency 1 0-3 0 ng/mL EC50
Catalog No. 50-003-32702 Supplier Creative Biomart Supplier No. THP0226
PDGF-BB isomer is found to be critical in angiogenesis and is highly expressed in tumor mediating oncogenic pericyte recruitment PDGF-BB is an important regulator of bone homeostasis repair and regeneration Potency 1 0-3 0 ng/mL EC50