Learn More
Genscript Corporation Antibody Protein Proteins Proteins 10 UG 1/EA
Supplier:  Genscript Corporation Z0372510
Encompass
Platelet-derived growth factor (PDGF) presenting in serum but absent from plasma was first discovered in an animal study by Lynch and co-workers in the late 1980s. It is a disulfide-linked dimer consisting of two peptides-chain A and chain B. PDGF has three subforms: PDGF-AA PDGF-BB and PDGF-AB. It is involved in many biological processes including hyperplasia embryonic neuron development chemotaxis and respiratory tubule epithelial cell development. The function of PDGF is mediated by two receptors (PDGFR-Alpha and PDGFR-beta ).
Catalog No. 50-252-9574