Learn More
ACROBiosystems ACROBiosystems Unconjugated Human PDGF-BB Protein, His,Avitag™
Catalog No. 501957273
Encompass_Preferred
Recombinant Protein;1MG;Unconjugated Human PDGF-BB, His,Avitag (PDB-H5127) is expressed from E.coli cells. It contains AA Ser 82 - Thr 190 (Accession # P01127-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 15.1 kDa. the protein migrates as 17 kDa under reducing (R) condition, and 35 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Unconjugated Human PDGF-BB, His,Avitag (Cat. No. PDB-H5127) at 5 µg/mL (100 µL/well) can bind Human PDGF R beta, Fc Tag (Cat. No. PDB-H5252) with a linear range of 0.02-1.2 µg/mL (QC tested).
Catalog No. 50-195-7273 Supplier ACROBiosystems Supplier No. PDBH51271MG
Recombinant Protein;1MG;Unconjugated Human PDGF-BB, His,Avitag (PDB-H5127) is expressed from E.coli cells. It contains AA Ser 82 - Thr 190 (Accession # P01127-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 15.1 kDa. the protein migrates as 17 kDa under reducing (R) condition, and 35 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Unconjugated Human PDGF-BB, His,Avitag (Cat. No. PDB-H5127) at 5 µg/mL (100 µL/well) can bind Human PDGF R beta, Fc Tag (Cat. No. PDB-H5252) with a linear range of 0.02-1.2 µg/mL (QC tested).