Learn More
Medchemexpress LLC PDGF-BB Human 100ug
SDP
Catalog No. 5000184929
Encompass_Preferred
Recombinant human PDGF-BB is a biologically active dimeric growth factor that binds PDGF receptors to stimulate cell proliferation, chemotaxis, extracellular matrix formation, vascularization, and tissue repair. Supplied as a lyophilized, tag-free powder produced in E. coli, it is intended for research applications studying PDGF signaling, osteogenesis, and tendon healing.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Demonstrated biological activity with ED50 ≤ 11 ng/mL in Balb/c 3T3 cell assay.
  • Approximately 14 kDa molecular weight (predicted 12.4 kDa).
  • Lyophilized from a 0.22 μm filtered buffer for stability and ease of reconstitution.
  • Stable at -20°C for two years; reconstituted stability at 4°C for one week.
  • Recommended to reconstitute at ≥100 μg/mL and to aliquot and freeze to avoid freeze-thaw.
Catalog No. 50-001-84929 Supplier Medchemexpress LLC Supplier No. HYP7055100UG
Recombinant human PDGF-BB is a biologically active dimeric growth factor that binds PDGF receptors to stimulate cell proliferation, chemotaxis, extracellular matrix formation, vascularization, and tissue repair. Supplied as a lyophilized, tag-free powder produced in E. coli, it is intended for research applications studying PDGF signaling, osteogenesis, and tendon healing.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Demonstrated biological activity with ED50 ≤ 11 ng/mL in Balb/c 3T3 cell assay.
  • Approximately 14 kDa molecular weight (predicted 12.4 kDa).
  • Lyophilized from a 0.22 μm filtered buffer for stability and ease of reconstitution.
  • Stable at -20°C for two years; reconstituted stability at 4°C for one week.
  • Recommended to reconstitute at ≥100 μg/mL and to aliquot and freeze to avoid freeze-thaw.