Learn More
Medchemexpress LLC HY-P7055 10ug Medchemexpress, PDGF-BB, Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP705510UG
Encompass_Preferred
Medchemexpress, HY-P7055 10ug PDGF-BB, Human CAS: Recombinant Human Platelet-derived Growth Factor-BB is the most active PDGF isoform, binds to PDGF receptor, promotes diverse cell types proliferation and osteogenesis, and further stimulates bone formation in fracture or defect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4300