PDGF-BB Protein, Human is a recombinant human platelet-derived growth factor BB that binds PDGF receptors and stimulates cell proliferation, chemotaxis, and osteogenesis. Supplied lyophilized in microgram quantities for research applications, the product includes formulation and storage guidance to support stability, reconstitution, and experimental use.
- Promotes proliferation and migration of diverse cell types.
- Supports bone regeneration and tendon healing experimental models.
- High purity (>95%) determined by reducing SDS-PAGE.
- Tag-free recombinant protein produced in E. coli expression system.
- Approximately 14 kDa molecular weight for experimental planning.
- Lyophilized with multiple buffer formulation options for reconstitution.
- Stable when stored frozen; aliquot and freeze for long-term storage.
- Reconstitution guidance recommends not reconstituting below 100 μg/mL in ddH2O.