Learn More
Medchemexpress LLC PDGF-BB Human His 10ug
SDP
Catalog No. 5000184542
Encompass_Preferred
Recombinant human PDGF-BB (platelet-derived growth factor-BB) is a dimeric growth factor produced in E. coli with an N-terminal 6xHis tag. It promotes cell proliferation, migration, and angiogenesis and is supplied lyophilized for research use. The product is high purity and formulated for stability; reconstitute per instructions and aliquot for long-term storage at low temperatures.
  • Recombinant his-tagged dimer expressed in E. coli.
  • Greater than 95% purity by reducing SDS-PAGE.
  • Lyophilized formulation for extended shelf life.
  • Recommended reconstitution at 100 μg/mL or higher.
  • Stable when aliquoted and stored at -20°C or -80°C.
  • Suitable for assays of cell proliferation, migration, and angiogenesis.
Catalog No. 50-001-84542 Supplier Medchemexpress LLC Supplier No. HYP7055B10UG
Recombinant human PDGF-BB (platelet-derived growth factor-BB) is a dimeric growth factor produced in E. coli with an N-terminal 6xHis tag. It promotes cell proliferation, migration, and angiogenesis and is supplied lyophilized for research use. The product is high purity and formulated for stability; reconstitute per instructions and aliquot for long-term storage at low temperatures.
  • Recombinant his-tagged dimer expressed in E. coli.
  • Greater than 95% purity by reducing SDS-PAGE.
  • Lyophilized formulation for extended shelf life.
  • Recommended reconstitution at 100 μg/mL or higher.
  • Stable when aliquoted and stored at -20°C or -80°C.
  • Suitable for assays of cell proliferation, migration, and angiogenesis.