Learn More
Medchemexpress LLC PDGF-BB Human His 2ug
SDP
Catalog No. 5000185506
Encompass_Preferred
Recombinant human PDGF-BB (His) is a lyophilized, N-terminal 6xHis-tagged growth factor expressed in E. coli and provided for research use in studies of cell signaling, proliferation, migration, and tissue regeneration. The protein is supplied with low endotoxin and high purity to support sensitive cell-based assays.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for cell assays.
  • Contains the mature sequence region S82-T190 with an N-terminal His tag.
  • Lyophilized formulation for extended shelf life and convenient storage.
  • Formulated with buffer components to enhance stability on reconstitution.
Catalog No. 50-001-85506 Supplier Medchemexpress LLC Supplier No. HYP7055B2UG
Recombinant human PDGF-BB (His) is a lyophilized, N-terminal 6xHis-tagged growth factor expressed in E. coli and provided for research use in studies of cell signaling, proliferation, migration, and tissue regeneration. The protein is supplied with low endotoxin and high purity to support sensitive cell-based assays.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for cell assays.
  • Contains the mature sequence region S82-T190 with an N-terminal His tag.
  • Lyophilized formulation for extended shelf life and convenient storage.
  • Formulated with buffer components to enhance stability on reconstitution.