Learn More
Medchemexpress LLC PDGF-BB Mouse 100ug
SDP
Catalog No. 5000197071
Encompass_Preferred
PDGF-BB Protein, Mouse is a recombinant PDGF-BB growth factor provided as a tag-free protein expressed in E. coli for research use in cell culture applications. The product is highly purified and displays the expected molecular weight by SDS-PAGE; it is intended for in vitro studies of cell signaling, proliferation, and migration. Store frozen and aliquot for long-term stability.
  • Recombinant, tag-free protein expressed in E. coli.
  • Purified to greater than 95% as determined by reducing SDS-PAGE.
  • Apparent molecular weight of approximately 13 kDa under reducing conditions.
  • Supplied in microgram-scale format suitable for cell culture research applications.
  • Stable when stored at -20°C; aliquot and keep at -80°C for extended storage.
  • For research use only; not for clinical use or sale to patients.
Catalog No. 50-001-97071 Supplier Medchemexpress LLC Supplier No. HYP7087100UG
PDGF-BB Protein, Mouse is a recombinant PDGF-BB growth factor provided as a tag-free protein expressed in E. coli for research use in cell culture applications. The product is highly purified and displays the expected molecular weight by SDS-PAGE; it is intended for in vitro studies of cell signaling, proliferation, and migration. Store frozen and aliquot for long-term stability.
  • Recombinant, tag-free protein expressed in E. coli.
  • Purified to greater than 95% as determined by reducing SDS-PAGE.
  • Apparent molecular weight of approximately 13 kDa under reducing conditions.
  • Supplied in microgram-scale format suitable for cell culture research applications.
  • Stable when stored at -20°C; aliquot and keep at -80°C for extended storage.
  • For research use only; not for clinical use or sale to patients.