Learn More
Medchemexpress LLC HY-P7087 10ug Medchemexpress, PDGF-BB, Mouse CAS: Purity:>98%
SDP
Catalog No. 501934327
Encompass_Preferred
Medchemexpress, HY-P7087 10ug PDGF-BB, Mouse CAS: Recombinant Mouse Platelet-derived Growth Factor-BB is an active PDGF isoform, used in cell culture applications. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4327 Supplier Medchemexpress LLC Supplier No. HYP708710UG
Medchemexpress, HY-P7087 10ug PDGF-BB, Mouse CAS: Recombinant Mouse Platelet-derived Growth Factor-BB is an active PDGF isoform, used in cell culture applications. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.