Learn More
Medchemexpress LLC PDGF-BB Mouse C-Hi 100ug
SDP
Catalog No. 5000184499
Encompass_Preferred
Recombinant mouse PDGF-BB (C-His) is a lyophilized, biologically active growth factor produced in E. coli and supplied as a high-purity powder for cell culture and signaling studies. It stimulates PDGF receptors to promote proliferation, migration, and survival pathways; store lyophilized at -20°C and freeze aliquots after reconstitution for long-term storage.
  • Recombinant mouse-derived PDGF-BB with C-terminal His tag.
  • High purity (≥98%) as assessed by SDS-PAGE.
  • Approximate molecular weight of ~15 kDa.
  • Lyophilized format for stable storage at -20°C.
  • Recommended reconstitution concentration ≥100 μg/mL; add carrier protein for long-term stability.
Catalog No. 50-001-84499 Supplier Medchemexpress LLC Supplier No. HYP70699A100UG
Recombinant mouse PDGF-BB (C-His) is a lyophilized, biologically active growth factor produced in E. coli and supplied as a high-purity powder for cell culture and signaling studies. It stimulates PDGF receptors to promote proliferation, migration, and survival pathways; store lyophilized at -20°C and freeze aliquots after reconstitution for long-term storage.
  • Recombinant mouse-derived PDGF-BB with C-terminal His tag.
  • High purity (≥98%) as assessed by SDS-PAGE.
  • Approximate molecular weight of ~15 kDa.
  • Lyophilized format for stable storage at -20°C.
  • Recommended reconstitution concentration ≥100 μg/mL; add carrier protein for long-term stability.