Learn More
Medchemexpress LLC PDGF-BB Mouse C-Hi 2ug
SDP
Catalog No. 5000185083
Encompass_Preferred
Recombinant mouse PDGF-BB (S82-T190) is an active growth factor produced in E. coli and supplied as a lyophilized powder with a C-terminal 6xHis tag. It is intended for cell signaling and cell culture applications, provided with high purity and detailed storage and handling recommendations.
  • High purity ≥98% as determined by SDS-PAGE.
  • Includes C-terminal 6xHis tag for purification or detection.
  • Lyophilized formulation for convenient storage and transport.
  • Recommend reconstitution to ≥100 μg/mL in 4 mM HCl.
  • Stable short term at 4°C after reconstitution; long-term storage at -20°C or -80°C recommended.
  • Approximately 15 kDa molecular weight by SDS-PAGE.
  • Suitable for cell signaling and tissue development research.
Catalog No. 50-001-85083 Supplier Medchemexpress LLC Supplier No. HYP70699A2UG
Recombinant mouse PDGF-BB (S82-T190) is an active growth factor produced in E. coli and supplied as a lyophilized powder with a C-terminal 6xHis tag. It is intended for cell signaling and cell culture applications, provided with high purity and detailed storage and handling recommendations.
  • High purity ≥98% as determined by SDS-PAGE.
  • Includes C-terminal 6xHis tag for purification or detection.
  • Lyophilized formulation for convenient storage and transport.
  • Recommend reconstitution to ≥100 μg/mL in 4 mM HCl.
  • Stable short term at 4°C after reconstitution; long-term storage at -20°C or -80°C recommended.
  • Approximately 15 kDa molecular weight by SDS-PAGE.
  • Suitable for cell signaling and tissue development research.