Learn More
Medchemexpress LLC PDGF-BB Mouse C-Hi 500ug
SDP
Catalog No. 5000184970
Encompass_Preferred
Recombinant mouse PDGF-BB with a C-terminal His tag, provided as a research-grade growth factor for in vitro studies. The protein is expressed in E. coli and purified for use in cell signaling, proliferation, and migration assays; users should refer to the manufacturer COA for batch-specific concentration, purity, formulation, and storage details.
  • Provides biologically active PDGF-BB for cell culture stimulation.
  • Facilitates reproducible signaling and proliferation assays.
  • C-terminal His tag enables straightforward purification and detection.
  • Expressed in a bacterial system for scalable production.
  • Batch certificate of analysis available for quality verification.
Catalog No. 50-001-84970 Supplier Medchemexpress LLC Supplier No. HYP70699A500UG
Recombinant mouse PDGF-BB with a C-terminal His tag, provided as a research-grade growth factor for in vitro studies. The protein is expressed in E. coli and purified for use in cell signaling, proliferation, and migration assays; users should refer to the manufacturer COA for batch-specific concentration, purity, formulation, and storage details.
  • Provides biologically active PDGF-BB for cell culture stimulation.
  • Facilitates reproducible signaling and proliferation assays.
  • C-terminal His tag enables straightforward purification and detection.
  • Expressed in a bacterial system for scalable production.
  • Batch certificate of analysis available for quality verification.