Learn More
Medchemexpress LLC PDGF-BB Mouse C-Hi 50ug
SDP
Catalog No. 5000184316
Encompass_Preferred
Recombinant mouse PDGF-BB with a C-terminal His tag, produced in E. coli and supplied lyophilized for use in cell biology and biochemical assays. The protein is quality-characterized for purity, molecular weight, bioactivity in cell proliferation assays, and low endotoxin to support reproducible in vitro studies. Datasheet and COA are available for batch-specific QC data.
  • Expressed in E. coli with a C-terminal 6xHis tag for detection and purification.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) measured by LAL assay.
  • Demonstrated bioactivity in cell proliferation assays (ED50 in the low ng/mL range).
  • Lyophilized formulation from 4 mM HCl for stability; recommended reconstitution to ≥100 μg/mL.
  • Supplied with downloadable datasheet and COA for batch verification.
Catalog No. 50-001-84316 Supplier Medchemexpress LLC Supplier No. HYP70699A50UG
Recombinant mouse PDGF-BB with a C-terminal His tag, produced in E. coli and supplied lyophilized for use in cell biology and biochemical assays. The protein is quality-characterized for purity, molecular weight, bioactivity in cell proliferation assays, and low endotoxin to support reproducible in vitro studies. Datasheet and COA are available for batch-specific QC data.
  • Expressed in E. coli with a C-terminal 6xHis tag for detection and purification.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) measured by LAL assay.
  • Demonstrated bioactivity in cell proliferation assays (ED50 in the low ng/mL range).
  • Lyophilized formulation from 4 mM HCl for stability; recommended reconstitution to ≥100 μg/mL.
  • Supplied with downloadable datasheet and COA for batch verification.