Learn More
Medchemexpress LLC PDGF-BB Mouse P.pa 10ug
SDP
Catalog No. 5000179836
Encompass_Preferred
PDGF-BB a crucial growth factor regulates diverse cellular processes including development proliferation migration survival and chemotaxis It is vital for tissue proliferation blood vessel development and kidney glomeruli formation PDGF-BB interacts with PDGFRA PDGFRB XLKD1 LRP1 and SORL1 highlighting its versatile cellular involvement PDGF-BB Protein Mouse (P pastoris His) is the recombinant mouse-derived PDGF-BB protein expressed by P pastoris with N-His labeled tag
Catalog No. 50-001-79836 Supplier Medchemexpress LLC Supplier No. HYP7335210UG
PDGF-BB a crucial growth factor regulates diverse cellular processes including development proliferation migration survival and chemotaxis It is vital for tissue proliferation blood vessel development and kidney glomeruli formation PDGF-BB interacts with PDGFRA PDGFRB XLKD1 LRP1 and SORL1 highlighting its versatile cellular involvement PDGF-BB Protein Mouse (P pastoris His) is the recombinant mouse-derived PDGF-BB protein expressed by P pastoris with N-His labeled tag