Learn More
Medchemexpress LLC PDGF-BB protein, cynomolgus (HEK293, mFc, solution) | 98.0%
SDP
Catalog No. 5000291007
Encompass_Preferred
Recombinant PDGF-BB (cynomolgus) is an mFc-tagged growth factor produced in HEK293 cells and supplied as a 0.22 μm-filtered PBS solution for research use. The protein is highly purified (>95% by reducing SDS-PAGE), shows apparent molecular weight shifts due to glycosylation, and is shipped frozen with recommended low-temperature storage to preserve activity.
  • mFc-tagged, mammalian-expressed protein suitable for receptor binding studies.
  • Supplied as a sterile, filtered PBS solution, ready for use.
  • High purity (>95%) as verified by reducing SDS-PAGE.
  • Stored and shipped frozen to maintain stability and activity.
  • Suitable for cell signaling, proliferation, and migration assays.
Catalog No. 50-002-91007 Supplier Medchemexpress LLC Supplier No. HYP75966A2UG
Recombinant PDGF-BB (cynomolgus) is an mFc-tagged growth factor produced in HEK293 cells and supplied as a 0.22 μm-filtered PBS solution for research use. The protein is highly purified (>95% by reducing SDS-PAGE), shows apparent molecular weight shifts due to glycosylation, and is shipped frozen with recommended low-temperature storage to preserve activity.
  • mFc-tagged, mammalian-expressed protein suitable for receptor binding studies.
  • Supplied as a sterile, filtered PBS solution, ready for use.
  • High purity (>95%) as verified by reducing SDS-PAGE.
  • Stored and shipped frozen to maintain stability and activity.
  • Suitable for cell signaling, proliferation, and migration assays.